Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ PTFE-Coated Steel Forceps
Stainless-steel forceps with Fluo-Kem™ PTFE FEP coated tips
$125.67 - $132.40
Specifications
Material Stainless Steel
Quantity (No. per Case) Each
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Product Type Price Quantity    
22261828
bel-art™
379370000
3.875 in. 98mm PTFE Coated entire length Pack of 2 Each for $132.40
Case of 24 Pack for $3,177.60
22-124768
bel-art™
H379350000
3.875 in. 98mm PTFE Coated 1.5 in.(38mm) Pack of 2 Each for $125.67
Case of 24 Pack for $3,016.08
22-107052
bel-art™
H379360000
3.875 in. 98mm PTFE Coated 3/4 in. (19mm) tip Pack of 2 Each for $125.67
Case of 24 Pack for $3,016.08
Description & Specifications
Specifications
Material Stainless Steel
Quantity (No. per Case) Each
  • Coating helps avoid scratches on delicate materials
  • Nonstick, corrosion- and chemical-resistant surface
  • Coating complies with FDA regulations for food contact