missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 10-13mm

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

Manufacturer:  Bel-Art™ F187500003

 View more versions of this product

Catalog No. 14-512-03




Description

Description

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable

Specifications

Specifications

72
Orange
8 x 4.4 x 2.5 in. (20.5 x 11.3 x 6.4cm)
6.4 cm
8.07 in.
Epoxy Coated Steel
11.3 cm
Autoclavable
13 mm
2.51 in.
10 to 13 mm Tubes
20.5 cm
4.45 in.
Test Tube Rack
SDS
Documents