Bel-Art™ SP Scienceware™ Poxygrid™ Petri Dish Dispensing Racks
Keep Petri dishes clean and orderly.
Manufacturer:  Bel-Art™ H189790000**
Catalog No. S67355
Description
Description
Epoxy-coated wire racks allow easy one-at-a-time dispensing of Petri dishes.
  • Load 3.9 × 0.5 in. (100 × 15mm) plastic Petri dishes from the top and slide out at the bottom
  • Available in two sizes
  • Single rack holds 14 Petri dishes; triple rack holds 42
Order Info
Not suitable for glass Petri dishes.
Specifications
Specifications
Not Autoclavable
11
5.75 x 12.75 x 11 in. (14.5 x 32 x 28cm)
5.75 in.
12.75 in.
11 in.
Steel Wire
10 x 2cm Petri dishes
14.5 cm
32 cm
28 cm