missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars

Center molded pivot ring minimizes friction and chattering

Manufacturer:  Bel-Art™ F371220035

 View more versions of this product

Catalog No. 03-410-525


 

Specifications Description & Specifications

Specifications

50
PTFE
Alnico Magnet
Polygon
1.37 in.
35mm
FDA, USP Class VI, ISO 9001:2008
White
Stir Bar
0.23 in.
6mm
Promotions

Promotions