Bel-Art™ SP Scienceware™ PiRack™ Pipetter Holder System
Mix and match components to suit your needs
Manufacturer:  Bel-Art™ F189540500
 View more versions of this product
Catalog No. 14-809-174
Description
Description
Polypropylene system is strong enough to support the weight of even the heaviest pipettor
  • Holds a single pipette with one clamp or add a triple clamp or extenders to increase the number of holders at a work location
  • Utility tray mounts between two clamps; ideal for storing pipetting supplies or other small items
  • Perfectly sized for No Wire™ Half Racks that hold tubes, vials, mini tubes and cuvettes (series 18748)
  • Attach to any shelf or benchtop edge up to 2.54cm (1 in.) thick using the large locking thumbscrew clamp
  • Supports many standard brands of pipettors
  • All components are chemical resistant and autoclavable at 121°C (250°F)
Specifications
Specifications
Autoclavable
Standard brands of pipettors
Polypropylene
Utility Tray
Documents