Bel-Art™ SP Scienceware™ Pinch Valves
Squeeze soft silicone body of the valve to permit flow and release pressure to suspend the flow, drip-free
Manufacturer:  Bel-Art™ 308480004
 View more versions of this product
Catalog No. 03-410-560
Specifications Description & Specifications
Specifications
Yes
10.18cm
Pinch Valve
Silicone, Rubber, Polypropylene
4.01 in.
Gravity flow, aspirator bottle