Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars in Colors
Octagonal shape for maximum surface area stirring
$9.51 - $773.77
Specifications
Coating PTFE
Material Magnet
Product Type Stir Bar
Shape Octagon
View More Specs
Products
Catalog Number Mfr. No. Case Quantity Color Diameter (English) Length (English) Price Quantity    
22-068106
bel-art™
371630000
20 Red, Yellow, Blue 0.31 in. 1 in. Pack of 6 Each for $106.71
22-261641
bel-art™
F371090010
50 Red 0.31 in. 1 in. Each for $16.47
Case of 50 Each for $823.50
22-261655
bel-art™
371090036
50 Blue 0.12 in. 0.5 in. Case of 50 Each for $773.77
22-261653
bel art products
371090034
Each for $9.51
22-261654
bel art products
371090035
50 Yellow 0.12 in. 0.5 in. Each for $15.48
Case of 50 Each for $774.00
Description & Specifications
Specifications
Coating PTFE
Material Magnet
Product Type Stir Bar
Shape Octagon
  • PTFE-coated
  • With pivot ring
  • Assorted sizes and colors