Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars
Octagon magnetic stirring bars with integral pivot ring
$12.48 - $47.39
Specifications
Autoclavable Yes
Case Quantity 50
Certifications/Compliance FDA, USP Class VI, ISO 9001:2008
Coating PTFE
Color White
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity  
19-120-176 Bel-Art™
F371101256
0.31 in. 8mm 0.5 in. 12.7mm Each for $13.23
Case of 50 Each for $661.50
22-066910 Bel-Art™
F371100078
0.31 in. 8mm 0.87 in. 22mm Each for $12.48
22-067074 Bel-Art™
F371100158
0.31 in. 8mm 1.62 in. 41.14mm Each for $18.13
22-067090 Bel-Art™
F371101238
0.37 in. 9.5mm 0.5 in. 12.7mm Each for $13.29
22-066969 Bel-Art™
F371100118
0.31 in. 8mm 1.12 in. 28.6mm Each for $13.23
22-067140 Bel-Art™
F371105838
0.37 in. 9.5mm 0.625 in. 16mm Each for $15.06
22-067157 Bel-Art™
F371107838
0.37 in. 9.5mm 0.87 in. 22mm Each for $15.58
22-067165 Bel-Art™
F371101388
0.37 in. 9.5mm 1.37 in. 35mm Each for $17.53
22-067173 Bel-Art™
F371100178
0.37 in. 9.5mm 1.87 in. 47.5mm Each for $23.53
22-067-181 Bel-Art™
F371102128
0.37 in. 9.5mm 2.5 in. 63.5mm Each for $28.45
Case of 50 Each for $1,422.50
22-271-841 Bel-Art™
F371100238
0.37 in. 9.5mm 2 in. 50.8mm Each for $24.58
Case of 50 Each for $1,229.00
22-271825 Bel-Art™
F371100138
0.37 in. 9.5mm 1 in. 25.4mm Each for $15.58
22-271833 Bel-Art™
F371101128
0.37 in. 9.5mm 1.5 in. 38mm Each for $18.48
Case of 50 Each for $924.00
22-271858 Bel-Art™
F371100212
0.31 in. 8mm 2.5 in. 63.5mm Each for $25.77
22-331488 Bel-Art™
F371100003
0.5 in. 12.7mm 3 in. 76.2mm Each for $47.39
Specifications
Description
  • Alnico magnet encapsulated in FDA grade, USP Class VI Teflon™ PTFE low-friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008 facility
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Yes
FDA, USP Class VI, ISO 9001:2008
White
Stir Bar
50
PTFE
Alnico Magnet
Octagon