Bel-Art™ SP Scienceware™ Octagon Magnetic Stirring Bar Assortments
Popular sizes and shapes
$123.02 - $250.30
Specifications
Case Quantity 6
Certifications/Compliance FDA
Coating PTFE
Color White
Product Type Stir Bar
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
22-331470 Bel-Art™
F371800000
2 micro and 12 standard bars two each of 0.5 x 0.31 in., 0.62 x 0.31 in., 1 x 0.37 in., 1.5 x 0.37 in., 2 x 0.37 in., 2.5 x 0.37 in. Each for $250.30
22-331462 Bel-Art™
371820000
12 standard bars two each of 0.5 x 0.31 in., 0.62 x 0.31 in., 1 x 0.37 in., 1.5 x 0.37 in., 2 x 0.37 in., 2.5 x 0.37 in. Each for $123.02
Specifications
Description
  • Convenient storage box segregates sizes and styles for easy, no-rummage organization
  • Octagonal spin bars, each with pivot ring
  • Made from Teflon™ PTFE
  • Autoclavable
Description & Specifications
Specifications
6
PTFE
Stir Bar
FDA
White
Octagon