missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ No-Wire™ Microcentrifuge Tube Racks

Open sides optimize use in crushed ice or water baths

$63.87 - $72.50

Specifications

Autoclavable Yes
Dimensions (L x W x H) 5.08 x 4.15 x 1.7 in. (12.8 x 10.5 x 4.3cm)
Material Polypropylene
Type Rack, Microcentrifuge Tube
Quantity 8/Cs.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Positions Holds Price Quantity  
Catalog Number Mfr. No. Positions Holds Price Quantity  
14-791-5G Bel-Art™
F188380005
63 0.5mL Tubes Each for $72.50
Case of 8 Each for $580.00
14-791-5H Bel-Art™
F188380015
42 1.5mL Tubes Each for $63.87
Case of 8 Each for $510.96
Specifications

Description

  • Molded polypropylene racks sink in water bath and will not tilt
  • Stackable compact size conserves valuable space
  • Autoclavable at 121°C (250°F)
  • Racks are shipped flat: easy-to-assemble

Order Info

Available in two sizes to accommodate most popular tube sizes. Dimensions (L x W x H): 5 × 4 × 1.7 in. (13 × 11 × 4.3cm)

Description & Specifications

Specifications

Yes
Polypropylene
8/Cs.
5.08 x 4.15 x 1.7 in. (12.8 x 10.5 x 4.3cm)
Rack, Microcentrifuge Tube