Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Modular Ultra-Low Five-Box Freezer Racks
Link together side-by-side or can be securely stacked for horizontal and vertical storage.
$30.13 - $99.50
Specifications
For Use With (Equipment) Ultra low freezers
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Color Product Type Price Quantity    
14-100-151
bel-art™
H189920000
68.9 x 14.9 x 1.7cm (27.12 x 5.87 x 0.68 in.) Each for $30.13
14-100-152
  Accessories
bel-art™
H189920002
68.6 x 15.2 x 8.9cm (27 x 6 x 3.5 in.) White Ultra Low Freezer Rack Each for $99.50
Case of 12 Each for $1,194.00
14-100-153
  Accessories
bel-art™
H189920012
27 x 6 x 3.5 in.; (68.6 x 15.2 x 8.9cm) Blue Ultra Low 5 Box Freezer Rack Each for $99.50
Case of 12 Each for $1,194.00
14-100-154
  Accessories
bel-art™
H189920022
68.6 x 15.2 x 8.9cm (27 x 6 x 3.5 in.) Yellow Ultra Low 5 Box Freezer Rack Each for $99.50
Case of 12 Each for $1,194.00
14-100-155
  Accessories
bel-art™
H189920032
68.6 x 15.2 x 8.9cm (27 x 6 x 3.5 in.) Green Ultra Low 5 Box Freezer Rack Each for $99.50
Case of 12 Each for $1,194.00
Description & Specifications
Specifications
For Use With (Equipment) Ultra low freezers
  • Configured to customize freezer space
  • Includes pull out drawer for easy sample retrieval even from back of freezer
  • Each lightweight, wire formed rack holds 5 freezer boxes up to 5.25 x 5.25 x 2 in. H
  • Available in 4 colors for easy sample identification
  • Top cover provides flat shelf space for additional storage