Bel-Art™ SP Scienceware™ Minipet™ Pipettors
The chemical resistance of PTFE resin in a self-filling, syringe-type repetitive dispenser
$1,480.38 - $1,480.38
Specifications
Accuracy ±0.5%
Autoclavable Autoclavable
Number of Channels 1
Includes PTFE 3-way check valve Teflon-coated plunger, ergonomic Polypropylene handle
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Increments Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Increments Price Quantity  
13-689-135A
View Documents Promotion Details
Bel-Art™
F379000010
1 mL 0.1mL
Each for $1,480.38
Description
Description
  • Features a proprietary PTFE 3-way valve and fittings on a borosilicate glass barrel
  • Self-locking PTFE plunger adjusts over full volume range of the syringe, yet won't move from original setting even after multiple dispensing
  • Safe for use with virtually any solvent or acid except HF
  • Autoclavable and suited for dispensing media and other sterile samples
  • Once the 18 in. long (20.3cm) PTFE filling tube is placed in a liquid reservoir, reagent flows through a closed system
  • Accuracy: ±0.5%
  • Molded plastic handle is ergonomically designed to minimize operator fatigue when used for extended periods
Order Info
Complete with filling tube and sinker.
Specifications
Specifications
±0.5%
Autoclavable
1
PTFE 3-way check valve Teflon-coated plunger, ergonomic Polypropylene handle