Bel-Art™ SP Scienceware™ Microtube Cutter
Safely, accurately and easily sections microcentrifuge tubes and eliminates the hazards of using razor blades or knives
Manufacturer:  Bel-Art™ F210180000
 View more versions of this product
Catalog No. 05-401A
Description
Description
  • Makes it easy to retrieve radioactive pellets from the tips of tubes without resuspension or the risk of splashed radioactive substances
  • Works well with polyethylene c or polypropylene microcentrifuge tubes up to 1.5mL
  • Chrome-plated body with replaceable steel blade
  • Maximum cut diameter is 9.4mm (0.370 in.)
Not recommended for polystyrene tubes.
Specifications
Specifications
Chrome-plated Steel
Microtube Cutter
Microtube cutter
Documents