Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Microcentrifuge Tube Ice Racks/Tray
Deep tray holds ice and water for extended use time
$57.13 - $136.37
Specifications
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Autoclavable Price Quantity    
01-816-24
bel-art™
H189050001
Yes Each for $136.37
Case of 12 Each for $1,636.44
11-394-459
bel-art™
189050011
No Each for $57.13
Description & Specifications
Specifications
Material Polypropylene
This tray with rack insert can accommodate up to fifty 1.5mL tubes (or other tubes with a diameter less than 12mm) for large-volume, temperature-stabilized benchtop processing
  • Includes flat perimeter rim for labeling tape
  • Can be safely stacked with identical racks when loaded or empty
  • Sturdy polypropylene provides long life and heavy usage
  • Steam autoclavable at 121°C (250°F)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.