Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Manomark™ Pens


Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth

Manufacturer: bel-art™  F133770000

 View more versions of this product

Catalog No. 13-385

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Product Type Manomark™ Pen
Quantity 12 Pack
Temperature (Metric) Range 0 to 155°C
Temperature (English) Range 32 to 310°F
For Use With (Application) Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth
Color Assorted