Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans
Keep temperature sensitive samples in an optimal environment. Bel Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans are the lightest icewares on the market.
$147.94 - $334.73
Specifications
For Use With (Application) Ideal for storing and transporting temperature sensitive samples in Ice, Alcohol Slurries, Water baths and more
Material Polymer Foam
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Color Dimensions (L x W x H) Temperature Range (Metric) Price Quantity    
11-675-129
bel-art™
188481102
1L Black 6 x 6 x 3 in. (15.2 x 15.2 x 7.6cm) -196 to 93°C Each for $147.94
11-676-38A
bel-art™
188484101
4L Blue 13 x 10 x 5.1 in. (33 x 26 x 13cm) -196 to 93°C Each for $270.55
11-675-123
bel-art™
188489101
9L Blue 16 x 13 x 5.1 in. (41 x 33 x 13cm) -196 to 93°C Each for $324.45
11-675-128
bel-art™
188481101
1L Blue 6 x 6 x 3 in. (15.2 x 15.2 x 7.6cm) -196 to 93°C Each for $147.94
11-675-58
bel-art™
188489102
9L Black 16 x 13 x 5.1 in. (41 x 33 x 13cm) -196 to 93°C Each for $334.73
11-676-38B
bel-art™
188484102
4L Black 33 x 25.4 x 12.7cm (13 x 10 x 5.1 in.) -196 to 93°C Each for $279.06
Description & Specifications
Specifications
For Use With (Application) Ideal for storing and transporting temperature sensitive samples in Ice, Alcohol Slurries, Water baths and more
Material Polymer Foam
  • Large enough to accommodate circular and rectangular floating test tube racks, flasks, bottles, etc.
  • Expanded polyvinyl chloride, therefore unbreakable under normal conditions
  • Withstand temperatures from -196° to +93°C (-321° to +200°F) without sweating or leaking
  • Will not absorb odors or moisture
  • Chemically resistant to highly acidic and basic solutions
  • Suitable for use with ice, dry ice alcohol slurries, or liquid nitrogen
  • Stackable
  • Wall thickness: 0.75 in. (2cm)