missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Magic Touch™ Iceware Ice Buckets

Lightweight ice buckets have excellent insulating properties for keeping temperature sensitive samples in an optimal environment

Manufacturer:  Bel-Art™ 188484002

 View more versions of this product

Catalog No. 11-676



 

Specifications

Description

Containers, Insulated; Magic Touch Icewares; Unbreakable; Expanded PVC with lid; Leak resistant; Excellent insulator for both hot and cold materials; Dimensions: 10 x 7 in. (25 x 18 cm); Capacity: 4L; Black

  • Temperature range: -196° to +100°C (-321° to +212°F)
  • Use with ice, dry ice, dry ice alcohol slurries, or liquid nitrogen
  • Nonsweating
  • Impervious to moisture and odors
  • Expanded polymer foam construction provides chemical resistance
  • Unbreakable under normal use conditions
  • Flared edge for easy carrying
  • Lid can be cut to make openings for thermometers, cooling coils, flasks, etc.
  • Available in two sizes: 2.5L buckets measure 22.2cm dia. at rim × 12.0cmH (8.75 × 4.75 in.); 4.0L buckets measure 21.5cm dia. at rim × 17.0cmH (8.5 × 6.7 in.)
  • Round floating racks available separately for 1.5mL and 15mL tubes

Description & Specifications

Specifications

Black
7 in.
Ice bucke with lid
PVC
-196 to 93°C
17.8cm
4L
Ideal for storing and transporting temperature sensitive samples in Ice, Alcohol Slurries, Water baths and more
-321 to 200°F
Promotions

Promotions