Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Magic Touch™ Iceware Ice Buckets
Lightweight ice buckets have excellent insulating properties for keeping temperature sensitive samples in an optimal environment
$216.94 - $269.48
Specifications
Product Type Ice bucke with lid
Material PVC
Temperature Range (English) -321 to 200°F
Temperature Range (Metric) -196 to 93°C
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Color Width (English) Price Quantity    
11-676-35
bel-art™
188484001
4L Blue Each for $269.48
11-676-70
bel-art™
188482001
2.5L Blue Each for $220.94
11-676-73
bel-art™
188482002
2.5L Black Each for $220.94
Case of 12 Each for $2,651.28
11-676
bel-art™
188484002
4L Black Each for $269.48
Case of 12 Each for $3,233.76
11-676-78
bel-art™
188482201
2.5L Blue Each for $216.94
11-676-79
bel-art™
188484201
4L Blue Each for $236.45
Description & Specifications
Specifications
Product Type Ice bucke with lid
Material PVC
Temperature Range (English) -321 to 200°F
Temperature Range (Metric) -196 to 93°C
  • Temperature range: -196° to +100°C (-321° to +212°F)
  • Use with ice, dry ice, dry ice alcohol slurries, or liquid nitrogen
  • Nonsweating
  • Impervious to moisture and odors
  • Expanded polymer foam construction provides chemical resistance
  • Unbreakable under normal use conditions
  • Flared edge for easy carrying
  • Lid can be cut to make openings for thermometers, cooling coils, flasks, etc.
  • Available in two sizes: 2.5L buckets measure 22.2cm dia. at rim × 12.0cmH (8.75 × 4.75 in.); 4.0L buckets measure 21.5cm dia. at rim × 17.0cmH (8.5 × 6.7 in.)
  • Round floating racks available separately for 1.5mL and 15mL tubes