missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Labeled Wash Bottles

Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.

Manufacturer:  Fisherbrand™ F116463832

 View more versions of this product

Catalog No. S90134


Alert:

All bottles are intended for dispensing use only, not for long-term storage. Use LYOB only with solvents compatible with LDPE.


Description

Description

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Toluene is red LDPE bottle
  • Polypropylene closure with leakproof design
  • Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol

Compliance

OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Specifications

Not Autoclavable
53 mm
1000 mL
Natural
LDPE,Polyethylene
Polypropylene
32 oz., 1000 mL
32 oz.
Universal
Natural
SDS
Documents