missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Lab Companion Cabinet Style Vacuum Desiccators, Clear

Clear desiccator provides vacuum sustainment in a space maximizing cube

Manufacturer:  Bel-Art™ F424004011

 View more versions of this product

Catalog No. 08-648-107


 

Specifications Description & Specifications

Specifications

Clear
15.62 in.
39.7cm
Hinged
13.62 in.
34.59cm
5kg
Cabinet
35L
10.8kg
14 in.
29.5cm
42cm
15.62 in.
15 in.
14.75 in.
37.46cm
14.75 x 14 x 13.62 in. (37.46 x 35.56 x 34.59cm)
13.6 in.
34.5cm
Desiccant tray, 3 perforated shelves, built-in ports, door-mounted analog vacuum gauge
3/5
1.2 cu. ft.
23.7 lb.
11.6 in.
16.5 in.
35.56cm
Vacuum Desiccator
39.67cm
38.1cm
Promotions

Promotions