Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Lab-Aire™ II Electric Dryers: Double-Sided, 120V


Electric-powered drying racks for glassware and plasticware

Manufacturer: bel-art™  F188190022

 View more versions of this product

Catalog No. 03-422-213

  • / Each
 

Description & Specifications

Specifications

Amps 8A
Autoclavable No
Includes 4 baskets, 38 pegs (12 small, 14 medium, 12 large) and 14 peg plugs
Tiers 2 Tier
Material ABS Plastic
Product Type Double Sided Electric Dryer
Voltage 120V