Bel-Art™ SP Scienceware™ Lab-Aire II™ Double-Sided Electric Benchtop Labware Dryers
Offers compact footprint and accelerates drying of labware with warm, circulating air.
$2,093.47 - $2,093.47
Specifications
Material ABS Plastic
Voltage 120V
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Tiers Product Type Price Quantity  
Catalog Number Mfr. No. Includes Tiers Product Type Price Quantity  
03-990-012 Bel-Art™
F188192303
4 baskets; 38 pegs 2 Tier Each for $2,093.47
Specifications
Description
Double-sided models double the capacity with only a slightly larger footprint and can be turned perpendicular to the benchtop for easy access to both sides.
  • 400w of drying power gently circulates heated air into and around items to decreases drying time
  • Heat selector knob with three temperature settings: high, medium and low
  • Power button illuminates when in use
  • 8 Amp at 120V, circuit breaker and independent thermo-fuse protects unit from overheating
  • Efficiency is enhanced with replaceable polyurethane foam filter
  • Redirect air flow by inserting plugs instead of pegs into unused peg holes
  • Sturdy bases with excellent balance; can be placed on any flat surface
  • Removable polypropylene baskets for storage of small labware and extra pegs
  • 120V
  • Two or three tier option
  • Baskets hold small glassware including test tubes, pipets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
ABS Plastic
120V