Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Jerricans
Feature a 20cm (8 in.) spout with a built-in air vent for smooth pouring
$136.97 - $298.97
Specifications
Closure Type Screw
Graduations Liters, U.S. gal.
Material HDPE
With Spout
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Depth (English) Height (English) With Closure Width (English) Price Quantity    
11-865-131
bel-art™
109350000
1.25 gal. 5.5 in. 10.6 in. 9.6 in. Each for $136.97
Case of 12 Each for $1,643.64
11-865-132
bel-art™
109360000
2.5 gal. 7.3 in. 16.2 in. 10.1 in. Each for $168.31
Case of 6 Each for $1,009.86
11-865-133
bel-art™
109370000
5 gal. 9.4 in. 16.2 in. 11.1 in. Each for $298.97
Case of 6 Each for $1,793.82
Description & Specifications
Specifications
Closure Type Screw
Graduations Liters, U.S. gal.
Material HDPE
With Spout
  • High-density polyethylene (HDPE) jerricans feature a large, smooth area for labeling
  • When not in use, the spout retracts into the threaded collar to conserve space
  • Collar is sealed by a leakproof screw cap
  • An extra hand grip on the bottom gives positive control when pouring
  • Graduated in U.S. gallons and liters, they have extra headspace for expansion of contents
  • The five gallon jerrican has a 2.5cm (1 in.) I.D. pouring spout; other sizes have 1.9cm (0.75 in.) I.D. spouts
  • The 5 gallon jerrican has a 2.5cm (1 in.) I.D. pouring spout; other sizes have 1.9cm (0.75 in.) I.D. spouts
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.