Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Inoculating Turntable
Ball bearings ensure smooth and effortless spinning for faster and more accurate inoculation
$340.84 - $340.84
Products
Catalog Number Mfr. No. Price Quantity    
14-512-50
bel-art™
409130000
Each for $340.84
Description & Specifications
  • This economical, hand-operated turntable allows even distribution of bacterial cells in Petri dishes from 60 through 100mm in diameter
  • The recessed, non-skid, neoprene rubber turntable top keeps the Petri dishes in place and is removable for cleaning
  • The body of the unit is rugged and easy-to-clean polypropylene for lightweight portability
  • Dimensions: 12.7 x 5cm H (5 x 2 in. H)
  • Weight: 0.4kg (14.3 oz.)