missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Hydrometer Racks

Safe storage for glass hydrometers

Manufacturer:  Bel-Art™ F189880000

 View more versions of this product

Catalog No. 14-792-16




Description

Description

  • Holds 18 hydrometers in vertical position
  • The middle and top plates have 25mm (1 in.) diameter holes, bottom plate wells taper from 25mm to 9mm to hold the hydrometer bulb securely
  • Extended base measures 21 x 21.6cm for tip-proof stability
  • Overall height: 25.1cm with handles for easy transport
  • Rugged polypropylene construction, steam autoclavable at 121°C (250°F)

Specifications

Specifications

Not Autoclavable
21.5 cm
2 Upper shelves with 1 in. dia. hole, 1 bottom shelf with tapered hole, 1 in. at top, 0.37 in. at bottom
18
20.6 cm
8.5 in.
Hydrometers (Long)
Polypropylene
8.25 in.
Hydrometer Rack
Documents