Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Hand-Held Colony Counter Accessories: Replacement Markers


For use with SP Scienceware™ Hand-Held Colony Counter

Manufacturer: bel-art™  H378620152

 View more versions of this product

Catalog No. 07-910-020

  • / Each
  • / Case of 10 Each
 

Description & Specifications

Specifications

Color Red
For Use With (Application) Works on Plastic or Glass
For Use With (Equipment) Scienceware™ colony counter
Product Type Replacement Felt Tip Pen