Bel-Art™ SP Scienceware™ Full-View Test Tube Supports
Versatile polypropylene racks feature pins for support or draining of test tubes in a compact space
$86.60 - $155.43
Specifications
Autoclavable Autoclavable
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Positions Price Quantity  
Catalog Number Mfr. No. Positions Price Quantity  
14-512-09
View Documents
Bel-Art™
H189110000
50
Each for $86.60
Case of 12 Each for $1,039.20
22-260644
View Documents
Bel-Art™
H189100189
160
Each for $155.43
Case of 24 Each for $3,730.32
Description
Description
  • Tubes can be stored upright, full or empty, or inverted over pins for draining
  • Steam autoclavable at 121°C (250°F)
Specifications
Specifications
Autoclavable
Polypropylene
SDS