Bel-Art™ SP Scienceware™ Flaskup™ Flask Holders
With slight taper on the inside wall which allows a variety of different size round bottom flasks to fit into one holder
Manufacturer:  Bel-Art™ F389512004
 View more versions of this product
Catalog No. 14-809-171C
Description
Description
  • Polypropylene make
  • Wide base stabilizes flasks to prevent spills
  • Side slots allow viewing of flask contents
  • Notches in sidewall accommodate tubing or instrumentation
  • Glass-filled resin adds weight for greater stability
  • Autoclavable at 121°C (250°F)
Specifications
Specifications
Autoclavable
50 mL Round and tapered bottom flasks
Polypropylene
Flask Stand
Documents