Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Fast-Release Pipette Pump™ Pipetters


Pipetting device with rapid release

Manufacturer: bel-art™  F379111002EMW

 View more versions of this product

Catalog No. S32281F

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Product Type Fast Release Pipet Filler
Autoclavable No
For Use With (Application) Pipets up to 2mL
For Use With (Equipment) 2mL
Material Polypropylene
Color Blue