Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Eye Wash Bottle


In an emergency, time matters. The SP Scienceware™ Eye Wash Bottle built-in eye cup provides immediate care.

Manufacturer: bel-art™  F248500000

 View more versions of this product

Catalog No. 10-028-5

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Product Type Eyewash Bottle
Capacity (English) 16 oz.
Color Natural
Capacity (Metric) 500mL
Includes Built-in eye cup, captive snap-on cap, Multilanguage directions
Material LDPE
For Use With (Application) Immediate emergency flushing of eye injuries
Capacity 16 oz. (500mL)