Bel-Art™ SP Scienceware™ Extracting-Nut Safe-Lab™ Stoppers
PTFE-resin stoppers with red polypropylene extractor nuts require no lubricants
Manufacturer:  Bel-Art™ F201971540
 View more versions of this product
Catalog No. 14-645-2F
Description
Description
  • Twist nut for removal—stopper lifts out easily without breaking glassware
  • Unaffected by boiling acids, hydrocarbons, ketones, esters, alcohols or alkalis, regardless of concentration
  • Nonfreezing, noncontaminating, chemically inert, easy to clean
  • Use at temperatures up to 210°C (410°F)
Specifications
Specifications
Includes: Red polypropylene extracting nut
34
210°C
Stopper
For glassware with ground joints
34/45
PTFE