Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Ergopet™ Memory Pipettors

State-of-the-art memory pipettor provides precise results every time.

Manufacturer: bel-art™  F379100013/EMD

 View all options for this product

Catalog No. S97507


Description & Specifications


Accuracy ±3%
Battery Type NiMH
Finger Style Ergonomic
Height (English) 15.5 in.
Height (Metric) 394mm

  • Aspirate and dispense precise amount of liquid repeatedly in “memory mode” without using graduations of pipette
  • Angled design and extended heights reduces muscle fatigue and repetitive stress injuries
  • Swiveling head of rotates up to 90° left or right, permitting an unobstructed view of meniscus and graduations and minimizing wrist strain
  • Soft silicone chuck accepts glass or plastic pipettes and rotates to change pipette angle from vertical to 15° outward
  • The hydrophobic filter within chuck protects pipettor from inadvertent backflow and can be easily replaced without tools
  • Comfortable grip and light weight make all Ergopet™ Pipettors feel balanced in hand, and provide comfortable strain-free operation
  • Available in range of heights