Bel-Art™ SP Scienceware™ Ergopet™ Memory Pipettors
State-of-the-art memory pipettor provides precise results every time.
$1,243.35 - $1,258.20
Specifications
Accuracy ±3%
Battery Type NiMH
Finger Style Ergonomic
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity  
S97506 Bel-Art™
F379100012/
11.5 in. 292mm Each for $1,258.20
S97505 Bel-Art™
F379100011/EMD
7.5 in. 191mm Each for $1,243.35
Specifications
Description
  • Aspirate and dispense precise amount of liquid repeatedly in “memory mode” without using graduations of pipette
  • Angled design and extended heights reduces muscle fatigue and repetitive stress injuries
  • Swiveling head of rotates up to 90° left or right, permitting an unobstructed view of meniscus and graduations and minimizing wrist strain
  • Soft silicone chuck accepts glass or plastic pipettes and rotates to change pipette angle from vertical to 15° outward
  • The hydrophobic filter within chuck protects pipettor from inadvertent backflow and can be easily replaced without tools
  • Comfortable grip and light weight make all Ergopet™ Pipettors feel balanced in hand, and provide comfortable strain-free operation
  • Available in range of heights
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
±3%
NiMH
Ergonomic