Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Ergopet™ Bellows Pipetters
A rugged bellows provides aspirating force without any need for batteries, power cords or vacuum tubing
$959.29 - $1,215.81
Specifications
Autoclavable No
For Use With (Application) Pipets up to 20mL
Product Type Bellows Pipetter
Material Plastic
View More Specs
Products
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity    
03-421-509
bel-art™
F379070012
15 in. 380mm Each for $1,116.23
03-421-510
bel-art™
F379070013
19 in. 480mm Each for $1,215.81
03-421-508
bel-art™
F379070011
11 in. 280mm Each for $959.29
Description & Specifications
Specifications
Autoclavable No
For Use With (Application) Pipets up to 20mL
Product Type Bellows Pipetter
Material Plastic
Features
  • Models with extended height raise the tip of the pipet up to 12 in. (30cm), dramatically lowering the hand position
  • Compressing the bellows provides about 100mL of pipetting volume
  • The tallest model places the pipet tip on a similar plane as the hand, increasing precision while placing tip into small vessels
  • Many users will be able to rest their elbow on the bench to support the pipetter
  • Soft silicone chuck accepts pipets from 1 to 20mL; simply press into the ribbed chuck for a leak-free seal
  • User-replaceable hydrophobic filter within chuck protects pipetter from overaspiration
  • Design reduces awkward wrist bending associated with standard pipetters
Design
  • Engineered to be lightweight while still feeling balanced in the hand; shortest model weighs only 201g; tallest, 277g
  • Eliminates hand and arm strain when working inside a hood or on the bench
  • Multiple ergonomic adjustments make pipetting motions better conform to your physiology and needs
  • Separate soft-touch aspiration and dispensing buttons are positioned to allow a comfortable, secure grip with easy reach for fine control
  • To keep the pipet vertical, nose or chuck of pipetter can be rotated to change angle of pipet from vertical to 15° outwards
  • Head can be positioned straight ahead in line with the hand or swung up to 90° left or right; allows unobstructed viewing of the pipetter while aspirating and dispensing
AVAILABLE ACCESSORIES
Wiire rack supports the shorter pipetter models vertically when not in use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.