Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Ergopet™ Air-Powered Pipettors
Light, ergonomic design that reduces awkward positioning, strain and fatigue to the pipetting hand and arm
$1,277.52 - $1,559.19
Specifications
Autoclavable No
Design Type Angled
Filter Type Hydrophobic
Grip Type Ergonomic
Voltage 120V
View More Specs
Products
Catalog Number Mfr. No. Height (English) Height (Metric) Product Type Price Quantity    
03-421-511
bel-art™
F379071011
8.26 in. 21cm Ergopet I Air Powered Pipetter Each for $1,277.52
03-421-512
bel-art™
F379071012
4 in. 10.1cm Ergopet II Air Powered Pipetter Each for $1,458.84
03-421-513
bel-art™
F379071013
8 in. 20.3cm Ergopet III Air Powered Pipetter Each for $1,559.19
Description & Specifications
Specifications
Autoclavable No
Design Type Angled
Filter Type Hydrophobic
Grip Type Ergonomic
Voltage 120V
Volume (Metric) 1 to 100mL
Warranty 1 Year
  • Feather-light weight and comfortable grip allows for hours of stress-free operation, tallest model weighs only 8.8 oz. (250g)
  • Extended handle lengths (4 or 8 in.) dramatically lower hand position allowing users to rest their elbow on bench while supporting their pipettor, especially when working in a hood with a lowered sash
  • Swiveling head can be positioned in line with the hand or turned up to 90° left or right to permit unobstructed viewing of meniscus and graduations while aspirating and dispensing
  • Chuck can be rotated to change angle of pipette from vertical to 15° outwards to ensure that meniscus is level at all times
  • Remote 120VAC air pump connected with 60 in. (1.5m) lightweight tubing provides continuous operation without needing battery charging
  • Soft silicone chuck accepts glass or plastic pipettes from 1-100mL with outside mouth diameter of 3.3 to 9.9mm
  • Hydrophobic filter within chuck can be replaced without using any tools and protects the pipette from inadvertent backflow
  • Not autoclavable
  • Store on optional accessory wire support stand
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.