Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells


Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket

Manufacturer: bel-art™  H402610000

 View more versions of this product

Catalog No. 08-666-16

  • / Each
 

Description & Specifications

Specifications

Quantity Each
Capacity (Metric) 5mL
Color Transparent
For Use With (Application) Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Material Acrylic
Product Type Dialysis Cell