Bel-Art™ SP Scienceware™ Ellipso-Spoon™ Spatula
Specially curved shape permits contact with all corners of bottles and containers to minimize sample loss
$73.20 - $178.63
Specifications
Autoclavable Yes
Product Type Ellipso-Spoon™ Sampler Spoon
Finish Polished
For Use With (Application) Designed to extracts crystals, granulated and solid substances, thick liquids and delicate materials
For Use With (Equipment) Bottles and containers
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
14-375-56 Bel-Art™
H368070021
8.26 in. 21cm Each for $81.10
Case of 12 Each for $973.20
14-375-55 Bel-Art™
H368060015
5.9 in. 15cm Each for $73.20
Case of 12 Each for $878.40
14-375-58 Bel-Art™
H368090040
15.74 in. 40cm Each for $178.63
Case of 12 Each for $2,143.56
Specifications
Description
  • Extracts crystals, granulated and solid substances, thick liquids, and delicate materials
  • Features a spoon end and a spatula end
  • Made of polished 304 grade stainless steel
  • Flameproof, autoclavable, and impervious to boiling water
Description & Specifications
Specifications
Yes
Polished
Bottles and containers
Curved
Ellipso-Spoon™ Sampler Spoon
Designed to extracts crystals, granulated and solid substances, thick liquids and delicate materials
Stainless Steel