missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Dry-Keeper™ Desiccator Cabinets

Clear, durable PVC desiccator cabinets have black ABS accents and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials

Manufacturer:  Bel-Art™ H420580001

 View more versions of this product

Catalog No. 08-647-28


 

Specifications

Description

Desiccator, Nonvacuum; Cabinet; Bel-Art; Horizontal; Molded clear and black methyl methacrylate; With hygrometer; Low RH (30-40%); 2 clear molded shelves, 2 removable; D 13.25 in. (34cm); 19.63L x 14.75W x 13.75 in. H (50 x 37 x 35cm)

 

  • Generous 2 cu. ft. interior volume
  • Adjustable, removable, and perforated shelves facilitate full circulation of air; additional shelves and rails sold separately
  • Large, in-door dial hygrometer for easy visualization of RH%
  • Gasketed door closes with two latches for a tight seal
  • Standard cabinets include desiccant tray (desiccant sold separately)
Horizontal Cabinets
  • Include two shelves
  • Shelf rails can be repositioned on any one of eight steps set at 1.1 in. (28mm) intervals
  • Interior opening measures 17.75 × 9 in. (45 × 23cm)
Vertical Cabinets
  • Includes three shelves
  • Shelf rails can be repositioned on any one of 15 steps set at 1.1 in. (28mm) intervals
  • Interior opening measures 10 × 17 in. (25 × 43cm)
Autodesiccators
  • 120VAC electronic dehumidification system maintains relative humidity levels at 30 to 40%†; keeps dry air circulating throughout the cabinet
  • Permanent desiccant contained in the control unit
  • Red light indicates operation of the 4.5 hour absorption period; green light indicates operation of the 0.5 hour regeneration of the desiccant

 

Description & Specifications

Specifications

Translucent
50cm
Adjusting dry storage space configuration for moisture-sensitive materials, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
78 cans
15.5 in.
2 cu. ft.
45cm
Methyl Methacrylate
34.2cm
19.62 in.
Hinged and gasketed
9.8 in.
Black ABS accents, 3 adjustable perforated shelves and shelf rails, desiccant tray and dial hygrometer, 2 metal hinges and latches, door-mounted hygrometer
39cm
17.7 in.
Horizontal
13.5 in.
2 adjustable perforated shelves
Promotions

Promotions