missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Drum and Carboy Funnels

Ensure optimum flow and volume.

Manufacturer:  Bel-Art™ H147120250

 View more versions of this product

Catalog No. 03-421-166


 

Specifications

Description

Funnel, Drum and Carboy; Bel-Art; SP Scienceware; Natural polypropylene plastic; Steam autoclavable; With handle at rim; Capacity: 4.3L; 30.9cm H; Stem O.D. x I.D. 5.2 x 2.3cm; Top O.D. x I.D.: 26 x 24.6cm; 9.1cm L stem

  • Molded of natural polypropylene plastic
  • Steam autoclavable at 121°C (250°F)
  • With handle at rim for added stability
Description & Specifications

Specifications

Yes
Optimum flow and volume
30.9cm
3.625 in.
2.0625 in.
4.3L (1.14 gal.)
Handle at rim
9.1cm
5.2cm
Polypropylene
Promotions

Promotions