Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Disposable Pipetting Reservoirs

Disposable and economical plastic pipetting reservoir. The SP Scienceware™ Disposable Pipetting Reservoirs includes ample clearance for all types of pipettors.

$210.76 - $257.88

Specifications

Dimensions (L x W x H) 6.12 x 3.62 x 1.25 in. (15.54 x 9.19 x 3.17cm)
Disposable Yes
Capacity (Metric) 100mL
Product Type Pipetting Reservoir
View More Specs
Products
Catalog Number Mfr. No. Packaging Sterile Price Quantity    

13-712-14

 
bel-art™
378770000
Bulk No Pack of 50 Each for $210.76

14-512-65

 
bel-art™
F378770001
Individually Wrapped Yes Pack of 50 Each for $257.88
Description & Specifications

Specifications

Dimensions (L x W x H) 6.12 x 3.62 x 1.25 in. (15.54 x 9.19 x 3.17cm)
Disposable Yes
Capacity (Metric) 100mL
Product Type Pipetting Reservoir

  • Ample clearance for all types of pipettors
  • Used for extended multichannel pipetting into microwell plates, Mini-Tubes, Cube 2ube™ Tubes and deep well blocks
  • 100ml capacity

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.