Bel-Art™ SP Scienceware™ Delrin™ Spring Tubing Clamps
Self-closing spring operated tubing clamp is suitable for all flexible tubing
Supplier: Bel-Art™ H182140000
Description
- Positive-actionclamp opens by squeezing handles together
- Naturally clamps shut to stop flow
- Made from chemical- and stain-resistant Delrin™ plastic and stainless steel
Specifications
Spring Style Tubing Clamp | |
Acetal Plastic, Stainless Steel | |
All flexible .46in. (1.16cm) O.D. tubing |