missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ CultureTemp 37°C Warming Plate for Cell Culture

Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells at an optimal 37°C (+/-1°C ) outside of the incubator

$696.21 - $789.31

Specifications

Depth (Metric) 25.4cm
Height (Metric) 5.7cm
Max. Temperature (Metric) 37°C
Wattage 22w
Width (Metric) 24.9cm
View More Specs

Products 2
Catalog Number Mfr. No. Voltage Price Quantity  
Catalog Number Mfr. No. Voltage Price Quantity  
14-513-613 View Documents Bel-Art™
F370150000
120V Each for $696.21
 
14-513-615 View Documents Bel-Art™
F370151000
220V Each for $789.31
 
Description

Description

  • Keep dishes outside the incubator longer as optimal temperature protects cell viability: No need to return to the incubator repeatedly
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Temperature remains uniform across the entire anodized aluminum warming plate that measures 17.8 D x 20.3cm W (7 x 8 in.)
  • Small footprint fits easily in a cell culture hood or on the workbench
  • UV-resistant polypropylene housing will not break down under germicidal UV lights
  • 15-minute countdown timer keeps track of time-sensitive applications
  • The unit is water resistant and safe for cleaning with ethanol or IPA

Compliance

UL approval pending.

Specifications

Specifications

25.4cm
37°C
24.9cm
17.8 x 20.3cm
5.7cm
22w
Anodized Aluminum