missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Compartmentalized Plastic Cases

Store small specimens or items.

Manufacturer:  Bel-Art™ F16632

 View more versions of this product

Catalog No. S47871A



Description

Description

  • Ideal for storing, organizing and displaying small specimens such as rocks, minerals and insects
  • Crystal-clear, styrene plastic affords full visibility of contents
  • Tight-fitting hinged covers protect the contents while keeping lab bench drawers neat and organized
  • Not recommended below 0°C
Specifications

Specifications

8.31 x 4.5 x 1.4 in. (21.1 x 11.4 x 3.6 cm)
8.313 in.
4.5 in.
3.6 cm
Plastic
11.4 cm
1.438 in.
Rectangular
Clear
21.1 cm
Compartment Tray
SDS
Documents
Product Certifications