Bel-Art™ SP Scienceware™ Compartmentalized Plastic Cases
Store small specimens or items.
$36.38 - $36.38
Specifications
Color Clear
Material Plastic
Type Compartment Tray
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity  
S47871A
View Documents
Bel-Art™
F16632
8.31 x 4.5 x 1.4 in. (21.1 x 11.4 x 3.6 cm)
Each for $36.38
Description
Description
  • Ideal for storing, organizing and displaying small specimens such as rocks, minerals and insects
  • Crystal-clear, styrene plastic affords full visibility of contents
  • Tight-fitting hinged covers protect the contents while keeping lab bench drawers neat and organized
  • Not recommended below 0°C
Specifications
Specifications
Clear
Plastic
Compartment Tray