Bel-Art™ Cloning Discs
Save time and reduce effort when isolating a large number of clones
Manufacturer: Bel-Art™ F378470003
Description
Save time and reduce effort when isolating a large number of clones
- The sterile paper discs transfer cells quickly and easily to a 24-well plate
- Cloning discs have been sterilized by Gamma radiation to be RNase and DNase free
Order Info
Supplied in vials of 50, two vials per pack.
Specifications
Paper | |
0.25 in. | |
Cloning Disc | |
Sterile |
RNase and DNase free | |
6.4mm | |
Ideal for cell transfer and propagation |