missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ Biohazard Disposal Bags with/without Warning Label
Economical, safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes
$110.22 - $313.27
Specifications
Product Type | Biohazard Bag |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | ASTM D1709-98 |
Disposable | Single-use |
For Use With (Application) | Disposal of used pipettes, petri dishes, culture plates, membrane filters and more |
Catalog Number | Mfr. No. | Color | Includes | Length (English) | Width (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Includes | Length (English) | Width (English) | Price | Quantity | ||
13-717-287
|
Bel-Art™
F131600005 |
Translucent | Non-printed without Sterilization Patch | 24 in. | 12 in. |
Pack of 100 Each for $110.22
|
|
||
13-717-289
|
Bel-Art™
F131600009 |
Clear | Biohazard Symbol with Sterilization Patch | 12 in. | 24 in. |
Pack of 100 Each for $115.68
|
|
||
13-717-288
|
Bel-Art™
F131610005 |
Translucent | Non-printed without Sterilization Patch | 30 in. | 24 in. |
Pack of 100 Each for $257.08
|
|
||
13-717-290
|
Bel-Art™
F131610009 |
Clear | Biohazard Symbol with Sterilization Patch | 24 in. | 30 in. |
Pack of 100 Each for $277.03
|
|
||
22-032926
|
Bel-Art™
F131620005 |
Translucent | Non-printed without Sterilization Patch | 36 in. | 24 in. |
Pack of 100 Each for $303.29
|
|
||
13-717-291
|
Bel-Art™
F131620009 |
Clear | Biohazard Symbol with Sterilization Patch | 24 in. | 36 in. |
Pack of 100 Each for $313.27
|
|
||
Description
Economical, safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes
- These transparent, polypropylene plastic biohazard bags are 0.03mm (1.5 mil) thick
- Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
- High temperature autoclavable at a maximum of 135°C (275°F)
- May also be used as conventional trash can liners
Specifications
Biohazard Bag | |
ASTM D1709-98 | |
Disposal of used pipettes, petri dishes, culture plates, membrane filters and more | |
1.5 mm |
Autoclavable | |
Single-use | |
Polypropylene |