missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Beta Box

Acrylic box with lid for excellent visibility of contents

$799.72 - $2,185.34

Specifications

Material Acrylic
Quantity Each
For Use With (Application) Radioactive Isotope Protection
Includes Non-Skid Feet, Removable Lid
View More Specs

Products 2
Catalog Number Mfr. No. Color Dimensions (L x W x H) Product Type Length (Metric) Interior Price Quantity  
Catalog Number Mfr. No. Color Dimensions (L x W x H) Product Type Length (Metric) Interior Price Quantity  
03-420-111
View Documents Promotion Details
Bel-Art™
F249830000
Clear 10 x 6 x 7 in. (25.4 x 15.2 x 17.9 cm) Beta Box 25.4 cm
Each for $799.72
 
03-420-112
View Documents Promotion Details
Bel-Art™
F249840000
Transparent 12 x 8 x 15 in. (30.4 x 20.3 x 38 cm) Beta box with Lid 30.48 cm
Each for $2,185.34
 
Description

Description

  • Made from 0.37 in. (9.5mm) clear acrylic
  • Includes removable lid and non-skid feet
  • Tight fitting joints reduce exposure to radioactive isotopes
  • L x W x H: 10 x 6 x 7 in. (25.4 x 15.2 x 17.9cm)

Specifications

Specifications

Acrylic
Each
Radioactive Isotope Protection
Non-Skid Feet, Removable Lid
SDS
Product Certifications