missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Autoradiography Pens

Easy marking of film with nontoxic and nonradioactive phosphorescent material

$210.06 - $220.09

Specifications

Color Bright Green
Includes Pocket clip and screw-knob advance to adjust writing tip
Length (English) 4.5 in.
Length (Metric) 114.3mm
Quantity Each
View More Specs

Includes: Pocket clip and screw-knob advance to adjust writing tip.


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
13-380-1A Bel-Art™
F133500000
Ideal for marking various films that are to be X-rayed with serial numbers, dates and exposure periods, film position markings Each for $210.06
Case of 6 Each for $1,260.36
13-380-1B Bel-Art™
F133510000
Ideal for marking radiography films like the Kodak XLS Each for $220.09
Case of 6 Each for $1,320.54
Specifications

Description

  • Solid phosphorescent material will not leak or dry out, (no bladder, vial or liquids to spill) Nonradioactive energy may be adjusted by exposure to white incandescent or fluorescent light
  • 3 to 4 minute energy emission leaves distinct impression without overexposing film
  • Bright green markings do not glow in the dark but are visible under normal conditions and on X-ray film
  • No Beta or other isotope emissions; Spectral peak, 5600Å
  • Screw knob advances writing tip; Pens are 4 1/2 in. long (11.4cm)
  • Not appropriate for phosphor screens

High Energy Level Markers
Greater energy emission strength and slightly longer energy emission duration
Suitable for marking X-Ray films similar to Kodak™ XLS
No Beta or other isotope emissions
Marking is unaffected by chilled films and an intensifier screen will not affect intensity
Not appropriate for phosphor screens

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Bright Green
4.5 in.
Each
Pocket clip and screw-knob advance to adjust writing tip
114.3mm