Bel-Art™ SP Scienceware™ Autoclavable Polypropylene Pails and Buckets
Rugged polypropylene pails with fitted lids/covers
$124.21 - $138.10
Specifications
Color White
Autoclavable Yes
Material Polypropylene
Product Type Pail
Handle Material Plated Steel
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Height (English) Height (Metric) Includes Price Quantity  
Catalog Number Mfr. No. Capacity Height (English) Height (Metric) Includes Price Quantity  
03-687-40 Bel-Art™
F167750000
1.72 gal., 7.6L 10 in. 25cm Removable support ring and steel plated bail handle, snug fitting cover Each for $138.10
Case of 6 Each for $828.60
03-687-41 Bel-Art™
F167760000
2.99 gal., 13.2L 12.75 in. 32cm Steel plated free swinging handle with grip, snug fitting cover Each for $124.21
Case of 6 Each for $745.26
Specifications
Description
  • Translucent polypropylene for easier viewing of fluid level
  • Steam autoclavable at 121°C (250°F)
  • 8-qt. pail has removable support ring and bail handle
  • 14-qt. pail has plated-steel bail handle with grip, no support ring
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
White
Polypropylene
Plated Steel
Yes
Pail