Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Autoclavable Polypropylene Carboys
Optimized for storing and dispensing large batches of culture media, distilled water and other solutions
$329.12 - $491.82
Specifications
Certifications/Compliance USP Class VI, 21CFR177.1520, FDA
Autoclavable Yes
Handles Two large
Material Polypropylene
For Use With (Application) For storing and dispensing large batches of culture media, distilled water and other solutions
View More Specs
Products
Catalog Number Mfr. No. Capacity Product Type Spigot Price Quantity    
01-189-0110
bel-art™
118460025
2.6 gal., 10L Carboy with polypropylene spigot Yes Each for $407.59
01-189-0111
bel-art™
118460050
5.3 gal., 20L Carboy with polypropylene spigot Yes Each for $491.82
03-410-805
bel-art™
F107940050
2.6 gal., 10L Carboy without polypropylene spigot No Each for $329.12
Description & Specifications
Specifications
Certifications/Compliance USP Class VI, 21CFR177.1520, FDA
Autoclavable Yes
Handles Two large
Material Polypropylene
For Use With (Application) For storing and dispensing large batches of culture media, distilled water and other solutions
  • These rugged, FDA-grade polypropylene carboys are 21 CFR 177.1520 compliant for direct contact with foods and beverages
  • USP Class VI certified for demands of biotechnology and pharmaceutical industries
  • Excellent chemical resistance
  • Large handles for ease of transport
  • All components are steam autoclavable at 121°C (250°F): when autoclaving, do not engage cap threads
  • The 83mm screw cap includes a built-in silicone gasket for positive seal
  • Molded-in bung accepts optional easy-to-use spigot and is located low on carboy for more complete drainage